Please login to your online account to display your discounted pricing

anti-KIR2DL3/CD158b2, APC, Clone: 180701, R&D Systems


Manufacturer: r & d systems  RDSFAB2014A100

 View more versions of this product

Catalog No. FAB2014A100

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Host Species Mouse
Format Purified
Quantity 100 Tests
Primary or Secondary Primary
Conjugate APC
Monoclonal or Polyclonal Monoclonal
Isotype IgG2a
Antigen KIR2DL3/CD158b2
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL3/CD158b2.
Cross Reactivity Human
Clone 180701
Purification Method Protein A or G purified from hybridoma culture supernatant