Please login to your online account to display your discounted pricing

anti-KIR2DL1/CD158a, PE, Clone: 143211, R&D Systems


Manufacturer: r & d systems  RDSFAB1844P100

 View more versions of this product

Catalog No. FAB1844P100

  • / Each
 

Description & Specifications

Specifications

Format Purified
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL1/CD158a.
Clone 143211
Quantity 100 Tests
Isotype IgG1
Cross Reactivity Human
Antigen KIR2DL1/CD158a
Host Species Mouse
Conjugate PE
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Purification Method Protein A or G purified from hybridoma culture supernatant