Please login to your online account to display your discounted pricing

anti-KIR2DL1/CD158a, Fluorescein, Clone: 143211, R&D Systems


Manufacturer: r & d systems  RDSFAB1844F100

 View more versions of this product

Catalog No. FAB1844F100

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL1/CD158a.
Clone 143211
Regulatory Status RUO
Format Purified
Antigen KIR2DL1/CD158a
Primary or Secondary Primary
Cross Reactivity Human
Conjugate Fluorescein
Isotype IgG1
Quantity 100 Tests
Purification Method Protein A or G purified from hybridoma culture supernatant
Monoclonal or Polyclonal Monoclonal