Please login to your online account to display your discounted pricing

anti-KIR2DL1/CD158a, APC, Clone: 143211, R&D Systems


Manufacturer: r & d systems  RDSFAB1844A025

 View more versions of this product

Catalog No. FAB1844A025

  • / Each
 

Description & Specifications

Specifications

Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL1/CD158a.
Quantity 25 Tests
Regulatory Status RUO
Clone 143211
Cross Reactivity Human
Antigen KIR2DL1/CD158a
Format Purified
Host Species Mouse
Isotype IgG1
Purification Method Protein A or G purified from hybridoma culture supernatant
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate APC