Please login to your online account to display your discounted pricing

anti-KIR2DL1/CD158a, Alexa Fluor 700, Clone: 143211, R&D Systems


Manufacturer: r & d systems  RDSFAB1844N025

 View more versions of this product

Catalog No. FAB1844N025

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Clone 143211
Quantity 25 Tests
Format Purified
Isotype IgG1
Antigen KIR2DL1/CD158a
Host Species Mouse
Primary or Secondary Primary
Conjugate Alexa Fluor 700
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL1/CD158a.
Purification Method Protein A or G purified from hybridoma culture supernatant
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human