Please login to your online account to display your discounted pricing

anti-KIR2DL1/CD158a, Alexa Fluor 700, Clone: 143211, R&D Systems


Manufacturer: r & d systems  RDSFAB1844N100

 View more versions of this product

Catalog No. FAB1844N100

  • / Each
 

Description & Specifications

Specifications

Antigen KIR2DL1/CD158a
Format Purified
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL1/CD158a.
Isotype IgG1
Quantity 100 Tests
Purification Method Protein A or G purified from hybridoma culture supernatant
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 143211
Conjugate Alexa Fluor 700
Cross Reactivity Human