Please login to your online account to display your discounted pricing

anti-Kininogen, Polyclonal, Invitrogen™


Manufacturer: pierce biotechnology inc  PA174131

Catalog No. PIPA174131

  • / Each
 

Description & Specifications

Specifications

Host Species Sheep
Quantity 10mg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Immunogen HMW Kininogen from human plasma.
Antigen Kininogen
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Format Purified
Isotype IgG