Please login to your online account to display your discounted pricing

anti-Ki-67/MKI67, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11090593SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11090593T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4288
Concentration 1mg/mL
Monoclonal or Polyclonal Polyclonal
Formulation Tris-citrate/phosphate, pH 7-8
Regulatory Status RUO
Gene Accession No. P46013
Conjugate Unlabeled
Primary or Secondary Primary
Format Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Synthetic peptide made to an internal portion of the human Ki67 protein (within residues 2100-2200). [Swiss-Prot No. P46013]
Antigen Ki-67/MKI67
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit
Quantity 0.025mL