Please login to your online account to display your discounted pricing

anti-Ki-67/MKI67, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11090593

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11090593

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-citrate/phosphate, pH 7-8
Primary or Secondary Primary
Gene ID (Entrez) 4288
Concentration 1mg/mL
Gene Accession No. P46013
Quantity 0.05mL
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Synthetic peptide made to an internal portion of the human Ki67 protein (within residues 2100-2200). [Swiss-Prot No. P46013]
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Ki-67/MKI67
Host Species Rabbit
Format Affinity Purified
Regulatory Status RUO