Please login to your online account to display your discounted pricing

anti-Ki-67/MKI67, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11090592SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11090592T

  • / Each
 

Description & Specifications

Specifications

Antigen Ki-67/MKI67
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. P46013
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human Ki67 protein (within residues 1200-1300). [Swiss-Prot No. P46013]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4288
Target Species Human
Target Species Murine