missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-KDM2A/FBXL11, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human KDM2A/FBXL11 protein (between residues 350-500) [UniProt Q9Y2K7]
Antigen KDM2A/FBXL11
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP178305SS Novus Biologicals Canada
NBP178305SS
0.025mL Each for $139.00
NBP178305 Novus Biologicals Canada
NBP178305
0.1mL Each for $309.05
Specifications Description & Specifications

Specifications

Rabbit
KDM2A/FBXL11
Affinity Purified
RUO
Q9Y2K7
PBS, 30% glycerol
A synthetic peptide made to an N-terminal portion of the human KDM2A/FBXL11 protein (between residues 350-500) [UniProt Q9Y2K7]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin)
22992
1.2mg/mL