Please login to your online account to display your discounted pricing

anti-KDM2A/FBXL11, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178305SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178305SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Host Species Rabbit
Gene ID (Entrez) 22992
Antigen KDM2A/FBXL11
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Gene Accession No. Q9Y2K7
Concentration 1.2mg/mL
Format Affinity Purified
Immunogen A synthetic peptide made to an N-terminal portion of the human KDM2A/FBXL11 protein (between residues 350-500) [UniProt Q9Y2K7]
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Quantity 0.025mL