Please login to your online account to display your discounted pricing

anti-KCNH1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142816SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142816SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene ID (Entrez) 3756
Test Specificity This antibody does not cross-react with Kv1.2. Human, mouse, rat, bovine and other vertebrates.
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Concentration 1mg/mL
Primary or Secondary Primary
Quantity 0.025mL
Host Species Rabbit
Format Affinity Purified
Immunogen Synthetic peptide made to an internal portion of rat KCNH1 (within residues 400-450).[Swiss-Prot No. Q63472]
Antigen KCNH1