Please login to your online account to display your discounted pricing

anti-KCNH1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142816

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142816

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3756
Test Specificity This antibody does not cross-react with Kv1.2. Human, mouse, rat, bovine and other vertebrates.
Concentration 1mg/mL
Antigen KCNH1
Quantity 0.1mL
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation PBS
Immunogen Synthetic peptide made to an internal portion of rat KCNH1 (within residues 400-450).[Swiss-Prot No. Q63472]
Regulatory Status RUO
Conjugate Unlabeled
Format Affinity Purified
Host Species Rabbit