Please login to your online account to display your discounted pricing

anti-KCNH1, Polyclonal, Novus Biologicals™

$354.00 - $354.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of rat KCNH1 (within residues 400-450).[Swiss-Prot No. Q63472]
Antigen KCNH1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP142816

 
novus biologicals canada
NBP142816
Unlabeled 0.1mL Each for $354.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of rat KCNH1 (within residues 400-450).[Swiss-Prot No. Q63472]
Antigen KCNH1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blot
Test Specificity This antibody does not cross-react with Kv1.2. Human, mouse, rat, bovine and other vertebrates.
Gene ID (Entrez) 3756
Formulation PBS
Concentration 1mg/mL