missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-JNK2, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 373-389 (KDQPSDAAVSSNATPSQ) of human JNK2 was used as immunogen; GenBank no. gi|21237736|ref|NP_002743.3|. This amino sequence is 100% homologous in cow and dog
Antigen JNK2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB6001297 Novus Biologicals Canada
NB6001297
0.1mg Each for $255.80
NB6001297SS Novus Biologicals Canada
NB6001297SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
JNK2
Purified
RUO
Cross-reacts with Human. Not yet tested in other species.
5601
0.5mg/mL
A synthetic peptide corresponding to amino acids 373-389 (KDQPSDAAVSSNATPSQ) of human JNK2 was used as immunogen; GenBank no. gi|21237736|ref|NP_002743.3|. This amino sequence is 100% homologous in cow and dog
Polyclonal
Primary
Western Blot
P45984
PBS containing 0.2% gelatin