Please login to your online account to display your discounted pricing

anti-JNK2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001297

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001297

  • / Each
 

Description & Specifications

Specifications

Antigen JNK2
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. P45984
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 373-389 (KDQPSDAAVSSNATPSQ) of human JNK2 was used as immunogen; GenBank no. gi|21237736|ref|NP_002743.3|. This amino sequence is 100% homologous in cow and dog
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5601
Test Specificity Cross-reacts with Human. Not yet tested in other species.
Target Species Human