Please login to your online account to display your discounted pricing

anti-JNK1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A recombinant protein made to the C-terminus (residues 228-451) of the C. elegans JNK1 alpha protein.
Antigen JNK1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001925

 
novus biologicals canada
NB1001925
Unlabeled 0.1mL Each for $309.05

NB1001925SS

 
novus biologicals canada
NB1001925SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A recombinant protein made to the C-terminus (residues 228-451) of the C. elegans JNK1 alpha protein.
Antigen JNK1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity C. elegans. This antibody does not appear to detect mammalian JNK protein.
Gene ID (Entrez) 5599