Please login to your online account to display your discounted pricing

anti-JMJD6/PSR, Polyclonal, Novus Biologicals™

$109.00 - $274.59

Specifications

Host Species Rabbit
Immunogen A peptide sequence corresponding to amino acids between 120 and 170 of human PTDSR was used as the immunogen. GenBank no NP_055982.
Antigen JMJD6/PSR
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224469

 
novus biologicals canada
NBP224469
Unlabeled 0.1mg Each for $274.59

NBP224469SS

 
novus biologicals canada
NBP224469SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A peptide sequence corresponding to amino acids between 120 and 170 of human PTDSR was used as the immunogen. GenBank no NP_055982.
Antigen JMJD6/PSR
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse, opossum, rat, dog, mouse and chimpanzee, 94% homologous in pig and cow, 83% homologous in xenopus and 77% homologous in chicken.
Gene Accession No. Q6NYC1
Gene ID (Entrez) 23210
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL