Please login to your online account to display your discounted pricing

anti-JMJD6/PSR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224469SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224469SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 23210
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q6NYC1
Primary or Secondary Primary
Quantity 0.025mg
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Primate
Concentration 0.5mg/mL
Immunogen A peptide sequence corresponding to amino acids between 120 and 170 of human PTDSR was used as the immunogen. GenBank no NP_055982.
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse, opossum, rat, dog, mouse and chimpanzee, 94% homologous in pig and cow, 83% homologous in xenopus and 77% homologous in chicken.
Antigen JMJD6/PSR
Host Species Rabbit
Format Affinity Purified
Applications Western Blotting