Please login to your online account to display your discounted pricing

anti-JMJD6/PSR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224469

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224469

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q6NYC1
Primary or Secondary Primary
Conjugate Unlabeled
Applications Western Blotting
Immunogen A peptide sequence corresponding to amino acids between 120 and 170 of human PTDSR was used as the immunogen. GenBank no NP_055982.
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse, opossum, rat, dog, mouse and chimpanzee, 94% homologous in pig and cow, 83% homologous in xenopus and 77% homologous in chicken.
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Primate
Formulation 100μg in 200μL PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 23210
Quantity 0.1mg
Regulatory Status RUO
Concentration 0.5mg/mL
Host Species Rabbit
Format Affinity Purified
Antigen JMJD6/PSR