Please login to your online account to display your discounted pricing

anti-JMJD2D, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP103357SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP103357SS

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Conjugate Unlabeled
Formulation Tris-citrate/phosphate, pH 7-8
Primary or Secondary Primary
Regulatory Status RUO
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human JMJD2D protein (within residues 460-520). [Swiss-Prot No. Q6B0I6]
Format Affinity Purified
Antigen JMJD2D
Test Specificity Human. Immunogen sequence has 92% homology to monkey.
Gene ID (Entrez) 55693
Gene Accession No. Q6B0I6
Applications Chromatin Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mL
Cross Reactivity Human