Please login to your online account to display your discounted pricing

anti-JMJD2D, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP103357

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP103357

  • / Each
 

Description & Specifications

Specifications

Antigen JMJD2D
Applications Chromatin Immunoprecipitation
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q6B0I6
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human JMJD2D protein (within residues 460-520). [Swiss-Prot No. Q6B0I6]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 55693
Test Specificity Human. Immunogen sequence has 92% homology to monkey.
Target Species Human