Please login to your online account to display your discounted pricing

anti-JMJD2D, Polyclonal, Novus Biologicals™

$111.78 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human JMJD2D protein (within residues 460-520). [Swiss-Prot No. Q6B0I6]
Antigen JMJD2D
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP103357

 
novus biologicals canada
NBP103357
Unlabeled 0.1mL Each for $309.05

NBP103357SS

 
novus biologicals canada
NBP103357SS
Unlabeled 0.025mL Each for $111.78
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human JMJD2D protein (within residues 460-520). [Swiss-Prot No. Q6B0I6]
Antigen JMJD2D
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Western Blotting
Test Specificity Human. Immunogen sequence has 92% homology to monkey.
Gene Accession No. Q6B0I6
Gene ID (Entrez) 55693
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL