Please login to your online account to display your discounted pricing

anti-JAMP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP236446

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP236446

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene Accession No. Q9P055
Primary or Secondary Primary
Conjugate Unlabeled
Antigen JAMP
Format Affinity Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mL
Concentration 1.0mg/mL
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51528
Cross Reactivity Human
Immunogen A synthetic peptide made to an internal portion of the human JAMP protein (between amino acids 20-150) [UniProt Q9P055]
Regulatory Status RUO
Host Species Rabbit