Please login to your online account to display your discounted pricing

anti-Jagged 2, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-210 of human JAG2 was used as the immunogen.
Antigen Jagged 2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227420

 
novus biologicals canada
NBP227420
Unlabeled 0.1mg Each for $307.00

NBP227420SS

 
novus biologicals canada
NBP227420SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-210 of human JAG2 was used as the immunogen.
Antigen Jagged 2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and monkey, 94% homologous in mouse, rat and horse, 76% homologous in opossum and 70% homologous in chicken.
Gene Accession No. Q9Y219
Gene ID (Entrez) 3714
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL