Please login to your online account to display your discounted pricing

IRP2 Polyclonal Antibody, Invitrogen™


IRP2 Antibody, PA1-16543, from Invitrogen™. Species Reactivity: Human, Mouse; Applications: Immunocytochemistry, Immunofluorescence, Western Blot.

Manufacturer: pierce biotechnology inc  PA116543

Catalog No. APPA6543

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 64602
Gene Symbols IREB2
Formulation 100μL purified antibody (1mg/mL) in tris glycine with 0.15M NaCl and 0.05% sodium azide
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Cross Reactivity Human
Cross Reactivity Mouse
Storage Requirements avoiding freeze/thaw cycles.
Isotype IgG
Species Reactivity Mouse
Gene Accession No. Q811J3
Gene Alias Iron-responsive element binding protein 2, IRE-BP 2, Iron regulatory protein 2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminal residues 1-100 of human IRP2 (UniProt P48200).
Quantity 100μL
Antigen IRP2
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Format Purified

IRP2 Antibody, PA1-16543, from Invitrogen. Species Reactivity: Human, Mouse; Applications: Immunocytochemistry, Immunofluorescence, Western Blot.

Iron regulatory protein 2 (IRP2) is a central post-transcriptional regulator of cellular and systemic iron metabolism. It is a cytosolic RNA-binding protein that binds to stem-loop structures. IRP2 is regulated by iron-mediated proteolysis. Hypoxia is important for normal tissue physiology as well as being a component of several pathophysiological conditions, including heart and cerebrovascular diseases and tumor growth. Because iron and oxygen are closely related, IRP2 also plays a role during hypoxia (low oxygen).
Host Species: Rabbit
Cross Reactivity: Human, Mouse
Isotype: IgG
Immunogen: Synthetic peptide corresponding to the N-terminal residues 1-100 of human IRP2 (UniProt P48200).
Formulation: 100μL purified antibody (1mg/mL) in tris glycine with 0.15M NaCl and 0.05% sodium azide
Synonyms: Iron-responsive element binding protein 2, IRE-BP 2, Iron regulatory protein 2
Uniprot Accession: Q811J3
Entrez GeneID: 64602
Gene Symbol: IREB2
This antibody is predicted to work in human based on 100% sequence homology, however preliminary data shows poor reactivity against human protein. This antibody also has 72% sequence homology with rat. Suggested positive control: 3T3 whole cell lysate.