Please login to your online account to display your discounted pricing

anti-IRP2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001798

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001798

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3658
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Mouse
Test Specificity Mouse. Preliminary results have shown poor reactivity against human protein. Immunogen has 94% homology with human, primate and rat proteins.
Concentration 1.15mg/mL
Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Host Species Rabbit
Quantity 0.1mL
Antigen IRP2
Format Affinity Purified
Immunogen A synthetic peptide made to an internal portion of the murine IRP2 protein sequence (between residues 100-200). [UniProt Q811J3]