Please login to your online account to display your discounted pricing

anti-IRP2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001797SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001797SS

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-glycine, 150mM NaCl
Conjugate Unlabeled
Quantity 0.025mL
Test Specificity Mouse. Predicted to work in human based on 100% sequence homology, however preliminary data shows poor reactivity against human protein. Immunogen has 72% homology to rat.
Gene ID (Entrez) 3658
Concentration 1mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen A synthetic peptide made to an N-terminal portion of the human IRP2 protein sequence (between residues 1-100). [UniProt P48200]
Cross Reactivity Mouse
Primary or Secondary Primary
Format Affinity Purified
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Antigen IRP2