Please login to your online account to display your discounted pricing

anti-IRP2, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human IRP2 protein sequence (between residues 1-100). [UniProt P48200]
Antigen IRP2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001797

 
novus biologicals canada
NB1001797
Unlabeled 0.1mL Each for $329.00

NB1001797SS

 
novus biologicals canada
NB1001797SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human IRP2 protein sequence (between residues 1-100). [UniProt P48200]
Antigen IRP2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Mouse. Predicted to work in human based on 100% sequence homology, however preliminary data shows poor reactivity against human protein. Immunogen has 72% homology to rat.
Gene ID (Entrez) 3658
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL