missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-IRE1 alpha, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within the human IRE1 alpha protein (within residues 200-300). [Swiss-Prot No.O75460]
Antigen IRE1 alpha
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB11059971 Novus Biologicals Canada
NB11059971
0.1mL Each for $309.05
NB11059971T Novus Biologicals Canada
NB11059971SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
IRE1 alpha
Affinity Purified
RUO
O75460
PBS, 30% glycerol
A synthetic peptide within the human IRE1 alpha protein (within residues 200-300). [Swiss-Prot No.O75460]
Polyclonal
Primary
Western Blot
2081
0.9mg/mL