Please login to your online account to display your discounted pricing

anti-IRE1 alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11059971SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11059971T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation PBS, 30% glycerol
Concentration 0.9mg/mL
Gene ID (Entrez) 2081
Gene Accession No. O75460
Immunogen A synthetic peptide within the human IRE1 alpha protein (within residues 200-300). [Swiss-Prot No.O75460]
Format Affinity Purified
Applications Western Blotting
Regulatory Status RUO
Quantity 0.025mL
Cross Reactivity Human
Cross Reactivity Mouse
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Primary or Secondary Primary
Antigen IRE1 alpha