Please login to your online account to display your discounted pricing

anti-IRE1 alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11059971

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11059971

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Gene Accession No. O75460
Concentration 0.9mg/mL
Host Species Rabbit
Primary or Secondary Primary
Conjugate Unlabeled
Gene ID (Entrez) 2081
Regulatory Status RUO
Applications Western Blotting
Immunogen A synthetic peptide within the human IRE1 alpha protein (within residues 200-300). [Swiss-Prot No.O75460]
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.1mL
Monoclonal or Polyclonal Polyclonal
Antigen IRE1 alpha