Please login to your online account to display your discounted pricing

anti-IRAK2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056447SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056447T

  • / Each
 

Description & Specifications

Specifications

Test Specificity In Jurkat, a 67 kDa band is observed.
Gene ID (Entrez) 3656
Concentration 0.5mg/mL
Gene Accession No. O43187
Conjugate Unlabeled
Formulation PBS containing 0.2% gelatin
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen This antibody was developed against a synthetic peptide of human IRAK-2.
Quantity 0.025mg
Format Purified
Antigen IRAK2
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting