Please login to your online account to display your discounted pricing

anti-IRAK2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056447

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056447

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O43187
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation PBS containing 0.2% gelatin
Test Specificity In Jurkat, a 67 kDa band is observed.
Concentration 0.5mg/mL
Gene ID (Entrez) 3656
Conjugate Unlabeled
Format Purified
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide of human IRAK-2.
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Antigen IRAK2
Quantity 0.1mg
Regulatory Status RUO