Please login to your online account to display your discounted pricing

anti-IRAK2, Polyclonal, Novus Biologicals™

$139.00 - $222.95

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide of human IRAK-2.
Antigen IRAK2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056447T

 
novus biologicals canada
NB10056447SS
Unlabeled 0.025mg Each for $139.00

NB10056447

 
novus biologicals canada
NB10056447
Unlabeled 0.1mg Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide of human IRAK-2.
Antigen IRAK2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In Jurkat, a 67 kDa band is observed.
Gene Accession No. O43187
Gene ID (Entrez) 3656
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL