missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Interleukin-8 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1427
Description
Anti-Interleukin-8 Antibody is an antibody against Interleukin-8 for use in ELISA and western blotting.Specifications
Interleukin-8 | |
Polyclonal | |
Liquid in PBS, no preservatives. | |
Rabbit | |
Protein A purfied | |
Primary | |
Human | |
Purified |
ELISA, Western Blot | |
Unconjugated | |
IL8; GCP-1; emoctakin; MDNCF; NAP1; CXCL8; AMCF-I; IL-8; Emoctakin; TSG-1; NAP-1; LYNAP; MONAP; SCYB8; GCP1; LUCT; LECT; K60; 3-10C; b-ENAP; NAF | |
Recombinant human IL-8. | |
100 μg | |
NM_000584.2 | |
2-8°C in undiluted aliquots for up to 12 months | |
IgG |
For Research Use Only