Please login to your online account to display your discounted pricing

anti-Integrin alpha M/CD11b, Clone: M1/70, R&D Systems

$139.08 - $139.08

Specifications

Antigen Integrin alpha M/CD11b
Clone M1/70
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MLDP5

 
r & d systems
RDSMLDP5
Unlabeled 1 Vial Each for $139.08
Description & Specifications

Specifications

Antigen Integrin alpha M/CD11b
Clone M1/70
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Regulatory Status RUO
Immunogen Con A-activated C57BL/10 splenocytes.
Isotype IgG2b
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary

Applications:

Cell Depletion