Please login to your online account to display your discounted pricing

anti-Integrin alpha M/CD11b, Clone: M1/70, R&D Systems


Manufacturer: r & d systems  RDSMLDP5

 View more versions of this product

Catalog No. MLDP5

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Quantity 1 Vial
Isotype IgG2b
Conjugate Unlabeled
Regulatory Status RUO
Clone M1/70
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Antigen Integrin alpha M/CD11b
Immunogen Con A-activated C57BL/10 splenocytes.
Host Species Rat
Cross Reactivity Mouse