Please login to your online account to display your discounted pricing

anti-Integrin β1, Polyclonal, EMD Millipore™


Manufacturer: emd millipore corporation  AB1952P

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AB1952PMI

  • / Each
 

Description & Specifications

Specifications

Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Storage Requirements 2-8°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Gene Symbols ITGB1; GPIIA; VLAB; OTTHUMP00000063732; MDF2; FNRB; OTTHUMP00000063731; OTTHUMP00000046253; CD29; OTTHUMP00000063733; MSK12
Gene ID (Entrez) NM_002211.2; NM_033666.1; NM_033667.1; NM_033668.1; NM_033669.1; NM_133376.1
Monoclonal or Polyclonal Polyclonal
Quantity 100μg
Purification Method Protein A purfied
Host Species Rabbit
Primary or Secondary Prrimary
Conjugate Unlabeled
Formulation Protein A Purified immunoglobulin. Liquid in PBS containing 0.1% sodium azide.
Immunogen Synthetic peptide corresponding to the C-terminus of human integrin beta 1 (a.a. CTTVVNPKYEGK)
Antigen Integrin β1