Please login to your online account to display your discounted pricing

anti-INSIG-1, Polyclonal, Novus Biologicals™

$144.63 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide representing an internal region of the human INSIG-1 protein (between residues 1-100) according to Swiss-Prot No. O15503.
Antigen INSIG-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11055244

 
novus biologicals canada
NB11055244
Unlabeled 0.1mL Each for $362.00

NB11055244T

 
novus biologicals canada
NB11055244SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide representing an internal region of the human INSIG-1 protein (between residues 1-100) according to Swiss-Prot No. O15503.
Antigen INSIG-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity This is specific for Insig1 but not Insig2.
Gene Accession No. O15503
Gene ID (Entrez) 3638
Formulation PBS, 30% glycerol
Concentration 1.02mg/mL