Please login to your online account to display your discounted pricing

anti-INHAT-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056619

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056619

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Primary or Secondary Primary
Test Specificity In HeLa, a 31 kDa band is observed. Reacts with Human, Chimpanzee, bovine and Rhesus Monkey. Predicted to react with Chicken, Mouse and Rat.
Gene Accession No. Q9Y3T9
Monoclonal or Polyclonal Polyclonal
Formulation PBS containing 0.1mg/mL BSA.
Conjugate Unlabeled
Quantity 0.2mL
Immunogen This antibody was raised against synthetic peptides of amino acids 87-103 and 231-246 of human INHAT-2.
Applications Western Blotting
Format Purified
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Host Species Rabbit
Antigen INHAT-2
Regulatory Status RUO