Please login to your online account to display your discounted pricing

anti-INHAT-2, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen This antibody was raised against synthetic peptides of amino acids 87-103 and 231-246 of human INHAT-2.
Antigen INHAT-2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056619

 
novus biologicals canada
NB10056619
Unlabeled 0.2mL Each for $257.41

NB10056619T

 
novus biologicals canada
NB10056619SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was raised against synthetic peptides of amino acids 87-103 and 231-246 of human INHAT-2.
Antigen INHAT-2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity In HeLa, a 31 kDa band is observed. Reacts with Human, Chimpanzee, bovine and Rhesus Monkey. Predicted to react with Chicken, Mouse and Rat.
Gene Accession No. Q9Y3T9
Formulation PBS containing 0.1mg/mL BSA.
Concentration 0.5mg/mL