Please login to your online account to display your discounted pricing

anti-INHAT-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056619SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056619T

  • / Each
 

Description & Specifications

Specifications

Antigen INHAT-2
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation PBS containing 0.1mg/mL BSA.
Gene Accession No. Q9Y3T9
Host Species Rabbit
Immunogen This antibody was raised against synthetic peptides of amino acids 87-103 and 231-246 of human INHAT-2.
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity In HeLa, a 31 kDa band is observed. Reacts with Human, Chimpanzee, bovine and Rhesus Monkey. Predicted to react with Chicken, Mouse and Rat.
Target Species Bovine
Target Species Human
Target Species Primate