Please login to your online account to display your discounted pricing

anti-Influenza A Haemagglutinin H1N1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 205-239 of influenza A H1N1 NS were used as the immunogen.
Antigen Influenza A Haemagglutinin H1N1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224938

 
novus biologicals canada
NBP224938
Unlabeled 0.1mg Each for $329.00

NBP224938SS

 
novus biologicals canada
NBP224938SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 205-239 of influenza A H1N1 NS were used as the immunogen.
Antigen Influenza A Haemagglutinin H1N1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Non-structural protein 1 from Influenza A virus (strain A/Swine/Iowa/15/1930 H1N1).
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL