Please login to your online account to display your discounted pricing

anti-Influenza A Haemagglutinin H1N1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 320-362 of human Influenza A H1N1 NA was used as the immunogen.
Antigen Influenza A Haemagglutinin H1N1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224937SS

 
novus biologicals canada
NBP224937SS
Unlabeled 0.025mg Each for $139.00

NBP224937

 
novus biologicals canada
NBP224937
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 320-362 of human Influenza A H1N1 NA was used as the immunogen.
Antigen Influenza A Haemagglutinin H1N1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL