Please login to your online account to display your discounted pricing

anti-Influenza A Haemagglutinin H1N1, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 238-270 of Influenza A H1N1 NA (swine flu) were used as the immunogen.
Antigen Influenza A Haemagglutinin H1N1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224956SS

 
novus biologicals canada
NBP224956SS
Unlabeled 0.025mg Each for $139.00

NBP224956

 
novus biologicals canada
NBP224956
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 238-270 of Influenza A H1N1 NA (swine flu) were used as the immunogen.
Antigen Influenza A Haemagglutinin H1N1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL