Please login to your online account to display your discounted pricing

anti-Importin-9, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056489SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056489T

  • / Each
 

Description & Specifications

Specifications

Antigen Importin-9
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. Q96P70
Host Species Rabbit
Immunogen A portion of amino acids 950-1000 of human Importin-9 was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 55705
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b and CRA_c), chimp, rhesus monkey, mouse (isoforms 1 and 2 and Importin-9a and Importin-9b), rat and bovine and 82% homologous in chicken. Cross reacts with Bovine, Chimpanzee, Human, Mouse, Rat and Rhesus Monkey. Predicted to react with Chicken.
Target Species Bovine
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat