Please login to your online account to display your discounted pricing

anti-IMP Dehydrogenase 2/IMPDH2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227069

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227069

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P12268
Gene ID (Entrez) 3615
Concentration 0.5mg/mL
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, platypus, common chimpanzee, orangutan, sumatran orangutan, panda, Carolina anole, guinea pig, new world monkey, dog, Chinese hamster, african bush elephant, crab-eating macaque, rhesus monkey, opossum, and white-cheeked gibbon, 94% in western clawed frog, zebra finch, pig, rabbit, wild turkey, chicken, horse, and cow, 83% in zebrafish, cichlid fish, and salmon.
Cross Reactivity Canine
Cross Reactivity Guinea Pig
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Primate
Cross Reactivity Rat
Immunogen Within the range of amino acids 122-162 from human IMPDH2 were used as the immunogen for this antibody.
Host Species Rabbit
Format Purified
Regulatory Status RUO
Applications Western Blotting
Antigen IMP Dehydrogenase 2/IMPDH2
Quantity 0.1mg