Please login to your online account to display your discounted pricing

anti-IMP Dehydrogenase 2/IMPDH2, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 122-162 from human IMPDH2 were used as the immunogen for this antibody.
Antigen IMP Dehydrogenase 2/IMPDH2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227069SS

 
novus biologicals canada
NBP227069SS
Unlabeled 0.025mg Each for $109.00

NBP227069

 
novus biologicals canada
NBP227069
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 122-162 from human IMPDH2 were used as the immunogen for this antibody.
Antigen IMP Dehydrogenase 2/IMPDH2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, platypus, common chimpanzee, orangutan, sumatran orangutan, panda, Carolina anole, guinea pig, new world monkey, dog, Chinese hamster, african bush elephant, crab-eating macaque, rhesus monkey, opossum, and white-cheeked gibbon, 94% in western clawed frog, zebra finch, pig, rabbit, wild turkey, chicken, horse, and cow, 83% in zebrafish, cichlid fish, and salmon.
Gene Accession No. P12268
Gene ID (Entrez) 3615
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL